RIE156326905 Rieke Metals, Inc.

3',5'-difluoro-3-(4-methoxyphenyl)propiophenone

  • MDL number MFCD03843191
  • Cas Number : 898776-43-7
  • Molecular Weight : 276.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-difluorophenyl)-3-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H14F2O2/c1-20-15-5-2-11(3-6-15)4-7-16(19)12-8-13(17)10-14(18)9-12/h2-3,5-6,8-10H,4,7H2,1H3
Inchi Key FOUKUWYGDYKPSM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0700-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-60
Availability details for
Quantity
  • Availability
  • Add to Cart