RIE156326907 Rieke Metals, Inc.

2',6'-dichloro-3-(3-methoxyphenyl)propiophenone

  • MDL number MFCD08064128
  • Cas Number : 898793-75-4
  • Molecular Weight : 309.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0700-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0700-62
Availability details for
Quantity
  • Availability
  • Add to Cart