RIE156326908 Rieke Metals, Inc.

Cyclopropyl 2-(4-methoxyphenyl)ethyl ketone

  • MDL number MFCD03843193
  • Cas Number : 87141-66-0
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0701-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0701-00
Availability details for
Quantity
  • Availability
  • Add to Cart