RIE156326911 Rieke Metals, Inc.

cyclohexyl 2-(4-methoxyphenyl)ethyl ketone

  • MDL number MFCD03843196
  • Cas Number : 885122-89-4
  • Molecular Weight : 246.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclohexyl-3-(4-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H22O2/c1-18-15-10-7-13(8-11-15)9-12-16(17)14-5-3-2-4-6-14/h7-8,10-11,14H,2-6,9,12H2,1H3
Inchi Key HCNALAOPYHNCIJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0704-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0704-00
Availability details for
Quantity
  • Availability
  • Add to Cart