RIE156326962 Rieke Metals, Inc.

cyclobutyl 2-(2-thiomethylphenyl)ethyl ketone

  • MDL number MFCD03843246
  • Cas Number : 898780-69-3
  • Molecular Weight : 234.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1402-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1402-00
Availability details for
Quantity
  • Availability
  • Add to Cart