RIE156326965 Rieke Metals, Inc.

3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843249
  • Cas Number : 96550-90-2
  • Molecular Weight : 256.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[4-(methylsulfanyl)phenyl]-1-phenyl-1-propanone
Inchi Code 1S/C16H16OS/c1-18-15-10-7-13(8-11-15)9-12-16(17)14-5-3-2-4-6-14/h2-8,10-11H,9,12H2,1H3
Inchi Key HKCJMVGYWDTQHC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1500-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-01
Availability details for
Quantity
  • Availability
  • Add to Cart