RIE156326967 Rieke Metals, Inc.

3'-methyl-2-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD07699941
  • Cas Number : 898780-77-3
  • Molecular Weight : 270.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-03
Availability details for
Quantity
  • Availability
  • Add to Cart