RIE156326973 Rieke Metals, Inc.

3'-cyano-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843254
  • Cas Number : 898780-89-7
  • Molecular Weight : 281.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-09
Availability details for
Quantity
  • Availability
  • Add to Cart