RIE156326977 Rieke Metals, Inc.

4'-carboethoxy-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843258
  • Cas Number : 898780-97-7
  • Molecular Weight : 328.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-{3-[4-(methylsulfanyl)phenyl]propanoyl}benzoate
Inchi Code 1S/C19H20O3S/c1-3-22-19(21)16-9-7-15(8-10-16)18(20)13-6-14-4-11-17(23-2)12-5-14/h4-5,7-12H,3,6,13H2,1-2H3
Inchi Key SVCYLMPRLYOMMC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1500-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-13
Availability details for
Quantity
  • Availability
  • Add to Cart