RIE156326981 Rieke Metals, Inc.

4'-bromo-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843262
  • Cas Number : 898781-06-1
  • Molecular Weight : 335.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-19
Availability details for
Quantity
  • Availability
  • Add to Cart