RIE156326985 Rieke Metals, Inc.

4'-fluoro-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843266
  • Cas Number : 898781-18-5
  • Molecular Weight : 274.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-23
Availability details for
Quantity
  • Availability
  • Add to Cart