RIE156326989 Rieke Metals, Inc.

2',6'-dimethyl-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843270
  • Cas Number : 898781-30-1
  • Molecular Weight : 284.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-dimethylphenyl)-3-[4-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C18H20OS/c1-13-5-4-6-14(2)18(13)17(19)12-9-15-7-10-16(20-3)11-8-15/h4-8,10-11H,9,12H2,1-3H3
Inchi Key MXZFCULUWUBTMB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1500-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-29
Availability details for
Quantity
  • Availability
  • Add to Cart