RIE156326994 Rieke Metals, Inc.

3'-chloro-4'-fluoro-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843275
  • Cas Number : 898781-45-8
  • Molecular Weight : 308.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-[4-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C16H14ClFOS/c1-20-13-6-2-11(3-7-13)4-9-16(19)12-5-8-15(18)14(17)10-12/h2-3,5-8,10H,4,9H2,1H3
Inchi Key DKJXPGSXEUOUHN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-1500-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-34
Availability details for
Quantity
  • Availability
  • Add to Cart