RIE156326997 Rieke Metals, Inc.

3-(4-thiomethylphenyl)-2'-trifluoromethylpropiophenone

  • MDL number MFCD03843278
  • Cas Number : 898781-53-8
  • Molecular Weight : 324.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-43
Availability details for
Quantity
  • Availability
  • Add to Cart