RIE156326999 Rieke Metals, Inc.

3-(4-thiomethylphenyl)-4'-trifluoromethylpropiophenone

  • MDL number MFCD03843280
  • Cas Number : 898781-57-2
  • Molecular Weight : 324.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-45
Availability details for
Quantity
  • Availability
  • Add to Cart