RIE156327001 Rieke Metals, Inc.

2'-chloro-4'-fluoro-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843282
  • Cas Number : 898781-61-8
  • Molecular Weight : 308.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-50
Availability details for
Quantity
  • Availability
  • Add to Cart