RIE156327003 Rieke Metals, Inc.

4'-chloro-2'-fluoro-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843284
  • Cas Number : 898781-65-2
  • Molecular Weight : 308.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-52
Availability details for
Quantity
  • Availability
  • Add to Cart