RIE156327007 Rieke Metals, Inc.

3',4'-dichloro-3-(4-thiomethylphenyl)propiophenone

  • MDL number MFCD03843271
  • Cas Number : 898781-73-2
  • Molecular Weight : 325.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1500-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1500-56
Availability details for
Quantity
  • Availability
  • Add to Cart