RIE156327015 Rieke Metals, Inc.

cyclobutyl 2-(4-thiomethylphenyl)ethyl ketone

  • MDL number MFCD03843294
  • Cas Number : 898781-89-0
  • Molecular Weight : 234.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1502-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1502-00
Availability details for
Quantity
  • Availability
  • Add to Cart