RIE156327016 Rieke Metals, Inc.

cyclopentyl 2-(4-thiomethylphenyl)ethyl ketone

  • MDL number MFCD03843295
  • Cas Number : 898781-91-4
  • Molecular Weight : 248.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-1503-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-1503-00
Availability details for
Quantity
  • Availability
  • Add to Cart