RIE156327018 Rieke Metals, Inc.

3-(3-chlorophenyl)-2'-methylpropiophenone

  • MDL number MFCD03843401
  • Cas Number : 898762-12-4
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-02
Availability details for
Quantity
  • Availability
  • Add to Cart