RIE156327019 Rieke Metals, Inc.

3-(3-chlorophenyl)-3'-methylpropiophenone

  • MDL number MFCD03843402
  • Cas Number : 898762-14-6
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-03
Availability details for
Quantity
  • Availability
  • Add to Cart