RIE156327022 Rieke Metals, Inc.

3-(3-chlorophenyl)-3'-methoxypropiophenone

  • MDL number MFCD03843405
  • Cas Number : 898762-23-7
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-06
Availability details for
Quantity
  • Availability
  • Add to Cart