RIE156327024 Rieke Metals, Inc.

3-(3-chlorophenyl)-2'-cyanopropiophenone

  • MDL number MFCD03843407
  • Cas Number : 898762-29-3
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[3-(3-chlorophenyl)propanoyl]benzonitrile
Inchi Code 1S/C16H12ClNO/c17-14-6-3-4-12(10-14)8-9-16(19)15-7-2-1-5-13(15)11-18/h1-7,10H,8-9H2
Inchi Key BJERNBTWZYRMSD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2000-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-08
Availability details for
Quantity
  • Availability
  • Add to Cart