RIE156327026 Rieke Metals, Inc.

3-(3-chlorophenyl)-4'-cyanopropiophenone

  • MDL number MFCD03843409
  • Cas Number : 898762-35-1
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-10
Availability details for
Quantity
  • Availability
  • Add to Cart