RIE156327027 Rieke Metals, Inc.

3-(3-chlorophenyl)-2'-carboethoxypropiophenone

  • MDL number MFCD03843410
  • Cas Number : 898762-38-4
  • Molecular Weight : 316.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[3-(3-chlorophenyl)propanoyl]benzoate
Inchi Code 1S/C18H17ClO3/c1-2-22-18(21)16-9-4-3-8-15(16)17(20)11-10-13-6-5-7-14(19)12-13/h3-9,12H,2,10-11H2,1H3
Inchi Key MSORHYXMDDKFRB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2000-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-11
Availability details for
Quantity
  • Availability
  • Add to Cart