RIE156327031 Rieke Metals, Inc.

3-(3-chlorophenyl)-4'-thiomethylpropiophenone

  • MDL number MFCD03843414
  • Cas Number : 898762-50-0
  • Molecular Weight : 290.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2000-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-15
Availability details for
Quantity
  • Availability
  • Add to Cart