RIE156327034 Rieke Metals, Inc.

3'-chloro-3-(3-chlorophenyl)propiophenone

  • MDL number MFCD03843417
  • Cas Number : 898762-59-9
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-20
Availability details for
Quantity
  • Availability
  • Add to Cart