RIE156327040 Rieke Metals, Inc.

3-(3-chlorophenyl)-2',5'-dimethylpropiophenone

  • MDL number MFCD03843423
  • Cas Number : 898787-07-0
  • Molecular Weight : 272.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-28
Availability details for
Quantity
  • Availability
  • Add to Cart