RIE156327045 Rieke Metals, Inc.

4'-chloro-3-(3-chlorophenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843428
  • Cas Number : 898787-18-3
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chloro-3-fluorophenyl)-3-(3-chlorophenyl)-1-propanone
Inchi Code 1S/C15H11Cl2FO/c16-12-3-1-2-10(8-12)4-7-15(19)11-5-6-13(17)14(18)9-11/h1-3,5-6,8-9H,4,7H2
Inchi Key RYADDEMAWSHCRH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2000-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-33
Availability details for
Quantity
  • Availability
  • Add to Cart