RIE156327050 Rieke Metals, Inc.

3-(3-chlorophenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD03843433
  • Cas Number : 898787-28-5
  • Molecular Weight : 312.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-chlorophenyl)-1-[3-(trifluoromethyl)phenyl]-1-propanone
Inchi Code 1S/C16H12ClF3O/c17-14-6-1-3-11(9-14)7-8-15(21)12-4-2-5-13(10-12)16(18,19)20/h1-6,9-10H,7-8H2
Inchi Key PNCKRMYKAODNFR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2000-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-44
Availability details for
Quantity
  • Availability
  • Add to Cart