RIE156327052 Rieke Metals, Inc.

4'-bromo-3-(3-chlorophenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843435
  • Cas Number : 898787-31-0
  • Molecular Weight : 341.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-49
Availability details for
Quantity
  • Availability
  • Add to Cart