RIE156327055 Rieke Metals, Inc.

4'-chloro-3-(3-chlorophenyl)-2'-fluoropropiophenone

  • MDL number MFCD03843438
  • Cas Number : 898787-34-3
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-52
Availability details for
Quantity
  • Availability
  • Add to Cart