RIE156327064 Rieke Metals, Inc.

3-(3-chlorophenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843447
  • Cas Number : 898787-48-9
  • Molecular Weight : 298.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2000-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2000-61
Availability details for
Quantity
  • Availability
  • Add to Cart