RIE156327071 Rieke Metals, Inc.

3-(4-chlorophenyl)-3'-methylpropiophenone

  • MDL number MFCD03843453
  • Cas Number : 898787-60-5
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-(3-methylphenyl)-1-propanone
Inchi Code 1S/C16H15ClO/c1-12-3-2-4-14(11-12)16(18)10-7-13-5-8-15(17)9-6-13/h2-6,8-9,11H,7,10H2,1H3
Inchi Key VFAOKBRFTYAYBF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-03
Availability details for
Quantity
  • Availability
  • Add to Cart