RIE156327072 Rieke Metals, Inc.

3-(4-chlorophenyl)-2'-methoxypropiophenone

  • MDL number MFCD03843455
  • Cas Number : 898787-62-7
  • Molecular Weight : 274.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-(2-methoxyphenyl)-1-propanone
Inchi Code 1S/C16H15ClO2/c1-19-16-5-3-2-4-14(16)15(18)11-8-12-6-9-13(17)10-7-12/h2-7,9-10H,8,11H2,1H3
Inchi Key WZZDQRGZXPOCNR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-05
Availability details for
Quantity
  • Availability
  • Add to Cart