RIE156327075 Rieke Metals, Inc.

3-(4-chlorophenyl)-3'-cyanopropiophenone

  • MDL number MFCD03843459
  • Cas Number : 898787-68-3
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-09
Availability details for
Quantity
  • Availability
  • Add to Cart