RIE156327079 Rieke Metals, Inc.

3-(4-chlorophenyl)-4'-carboethoxypropiophenone

  • MDL number MFCD03843463
  • Cas Number : 898787-76-3
  • Molecular Weight : 316.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[3-(4-chlorophenyl)propanoyl]benzoate
Inchi Code 1S/C18H17ClO3/c1-2-22-18(21)15-8-6-14(7-9-15)17(20)12-5-13-3-10-16(19)11-4-13/h3-4,6-11H,2,5,12H2,1H3
Inchi Key LJTOYEWGSIDVCG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-13
Availability details for
Quantity
  • Availability
  • Add to Cart