RIE156327083 Rieke Metals, Inc.

4'-bromo-3-(4-chlorophenyl)propiophenone

  • MDL number MFCD03843467
  • Cas Number : 296271-45-9
  • Molecular Weight : 323.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-19
Availability details for
Quantity
  • Availability
  • Add to Cart