RIE156327085 Rieke Metals, Inc.

3-(4-chlorophenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843470
  • Cas Number : 898787-91-2
  • Molecular Weight : 262.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-(3-fluorophenyl)-1-propanone
Inchi Code 1S/C15H12ClFO/c16-13-7-4-11(5-8-13)6-9-15(18)12-2-1-3-14(17)10-12/h1-5,7-8,10H,6,9H2
Inchi Key CVFTUTZFBZCOBY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-22
Availability details for
Quantity
  • Availability
  • Add to Cart