RIE156327086 Rieke Metals, Inc.

3-(4-chlorophenyl)-4'-fluoropropiophenone

  • MDL number MFCD03843471
  • Cas Number : 56201-99-1
  • Molecular Weight : 262.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-23
Availability details for
Quantity
  • Availability
  • Add to Cart