RIE156327088 Rieke Metals, Inc.

3-(4-chlorophenyl)-2',4'-dimethylpropiophenone

  • MDL number MFCD03843473
  • Cas Number : 898787-97-8
  • Molecular Weight : 272.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-27
Availability details for
Quantity
  • Availability
  • Add to Cart