RIE156327091 Rieke Metals, Inc.

3-(4-chlorophenyl)-3',5'-dimethylpropiophenone

  • MDL number MFCD03843477
  • Cas Number : 898788-09-5
  • Molecular Weight : 272.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-(3,5-dimethylphenyl)-1-propanone
Inchi Code 1S/C17H17ClO/c1-12-9-13(2)11-15(10-12)17(19)8-5-14-3-6-16(18)7-4-14/h3-4,6-7,9-11H,5,8H2,1-2H3
Inchi Key YXRANZUQENNUSQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-31
Availability details for
Quantity
  • Availability
  • Add to Cart