RIE156327093 Rieke Metals, Inc.

4'-Chloro-3-(4-chlorophenyl)-3'-fluoropropiophenone

  • MDL number MFCD03843479
  • Cas Number : 898788-15-3
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-33
Availability details for
Quantity
  • Availability
  • Add to Cart