RIE156327095 Rieke Metals, Inc.

2'-chloro-3-(4-chlorophenyl)propiophenone

  • MDL number MFCD03843481
  • Cas Number : 898788-21-1
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-36
Availability details for
Quantity
  • Availability
  • Add to Cart