RIE156327102 Rieke Metals, Inc.

3'-chloro-3-(4-chlorophenyl)-5'-fluoropropiophenone

  • MDL number MFCD03843488
  • Cas Number : 898788-35-7
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-51
Availability details for
Quantity
  • Availability
  • Add to Cart