RIE156327104 Rieke Metals, Inc.

3-(4-chlorophenyl)-2',3'-dichloropropiophenone

  • MDL number MFCD03843490
  • Cas Number : 898788-39-1
  • Molecular Weight : 313.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorophenyl)-1-(2,3-dichlorophenyl)-1-propanone
Inchi Code 1S/C15H11Cl3O/c16-11-7-4-10(5-8-11)6-9-14(19)12-2-1-3-13(17)15(12)18/h1-5,7-8H,6,9H2
Inchi Key HZYRRJTYQUEAAR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-2100-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-53
Availability details for
Quantity
  • Availability
  • Add to Cart