RIE156327106 Rieke Metals, Inc.

3-(4-chlorophenyl)-2',5'-dichloropropiophenone

  • MDL number MFCD03843492
  • Cas Number : 898788-43-7
  • Molecular Weight : 313.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-55
Availability details for
Quantity
  • Availability
  • Add to Cart