RIE156327108 Rieke Metals, Inc.

3-(4-chlorophenyl)-3',5'-dichloropropiophenone

  • MDL number MFCD03843494
  • Cas Number : 898788-45-9
  • Molecular Weight : 313.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-57
Availability details for
Quantity
  • Availability
  • Add to Cart