RIE156327110 Rieke Metals, Inc.

3-(4-chlorophenyl)-3',4'-difluoropropiophenone

  • MDL number MFCD03843496
  • Cas Number : 898788-47-1
  • Molecular Weight : 280.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-2100-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-2100-59
Availability details for
Quantity
  • Availability
  • Add to Cart